Product Details
- SNP ID
-
rs6321142
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:31001043 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATTGCATATTGAAGATAAACAAT[A/C]TCATCAAGTTATAGATTTTAGGGTT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Rit2
PubMed Links
Gene Details
- Gene
- Rit2
- Gene Name
- Ras-like without CAAX 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_009065.2 |
|
Intron |
|
|
NP_033091.1 |
View Full Product Details