Product Details

SNP ID
rs3023440
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:91646020 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTTTACTTTGGGATTACATATTTAA[A/G]GAAAACCCAAGCAGACTGGTAAGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Gart PubMed Links

Gene Details

Gene
Gart
Gene Name
phosphoribosylglycinamide formyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_010256.2 Intron NP_034386.2
Gene
Son
Gene Name
Son DNA binding protein
There are no transcripts associated with this gene.

View Full Product Details