Product Details

SNP ID
rs13482495
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:27814927 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCTCCCTGCCTCAGGAGAGGT[A/G]CTCATGGCGGTGAGGTAAGACTCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Trio PubMed Links

Gene Details

Gene
Trio
Gene Name
triple functional domain (PTPRF interacting)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081302.1 Intron NP_001074771.1

View Full Product Details