Product Details
- SNP ID
-
rs3659971
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:86917832 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCTGGGTTATGTCCATAATGACA[T/C]CAACAGAGTAGTTCTTTGGGAGACC
- Phenotype
-
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Acer2
PubMed Links
Gene Details
- Gene
- Acer2
- Gene Name
- alkaline ceramidase 2
There are no transcripts associated with this gene.
View Full Product Details