Product Details
- SNP ID
-
rs4228731
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:55394554 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAGACTTGTCAAGCTCTTCACAT[C/T]GCTGTGAGAGTTTGCATGGTTAATG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Sparc
PubMed Links
Gene Details
- Gene
- Sparc
- Gene Name
- secreted acidic cysteine rich glycoprotein
There are no transcripts associated with this gene.
View Full Product Details