Product Details

SNP ID
rs13481737
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:29304963 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGCTCTAGACCCAGCACTCTCCTAT[A/G]AAAACACATATAAACTAATCAAATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Cdkal1 PubMed Links

Gene Details

Gene
Cdkal1
Gene Name
CDK5 regulatory subunit associated protein 1-like 1
There are no transcripts associated with this gene.

View Full Product Details