Product Details

SNP ID
rs13476928
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:174095065 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TACTGTCTCTCATTACTCAGGGAAC[A/G]GTCCTTGATAGAATTGACTATAATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Stx16 PubMed Links

Gene Details

Gene
Stx16
Gene Name
syntaxin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102423.1 808 Silent Mutation NP_001095893.1
NM_001102424.1 808 Silent Mutation NP_001095894.1
NM_001102425.1 808 Silent Mutation NP_001095895.1
NM_172675.4 808 Silent Mutation NP_766263.2

View Full Product Details