Product Details

SNP ID
rs13476812
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:142899466 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CACAGATCCTACCTGCCTGCCTACT[C/G]AAATCATAAGGCTTCTGTAATGGTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Kif16b PubMed Links

Gene Details

Gene
Kif16b
Gene Name
kinesin family member 16B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081133.2 Intron NP_001074602.1
Gene
Kif16bos
Gene Name
kinesin family member 16B, opposite strand
There are no transcripts associated with this gene.

View Full Product Details