Product Details
- SNP ID
-
rs4221305
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:96722862 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAATCTAGGAATTGGCTATTTCAG[A/G]GCCCTTTTTGTACATCACATCAAAC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Dscam
PubMed Links
Gene Details
- Gene
- Dscam
- Gene Name
- Down syndrome cell adhesion molecule
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031174.4 |
|
Intron |
|
|
NP_112451.1 |
View Full Product Details