Product Details

SNP ID
rs13481078
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:67186091 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACAGGTTGCACAACACAACTTTTAT[C/T]ACTTGAAGTTATGCTTATTTTTTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Myh2 PubMed Links

Gene Details

Gene
Myh2
Gene Name
myosin, heavy polypeptide 2, skeletal muscle, adult
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039545.2 Intron NP_001034634.2

View Full Product Details