Product Details

SNP ID
rs13477174
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.3:68572628 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTTTGGGGTCTGCGTCCTAGGATGT[A/T]CTGAGATGGTACATCAGGAGAACTG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Gm21949 PubMed Links

Gene Details

Gene
Gm21949
Gene Name
predicted gene, 21949
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113419.2 261 Intron NP_001106890.1
Gene
Schip1
Gene Name
schwannomin interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113420.1 261 Intron NP_001106891.1
NM_001113421.1 261 Intron NP_001106892.1
NM_001282044.1 261 Intron NP_001268973.1
NM_001282045.1 261 Intron NP_001268974.1
NM_013928.5 261 UTR 5 NP_038956.2

View Full Product Details