Product Details

SNP ID
rs3718901
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:43243301 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGCTAAAACTCATACAATTACACAG[G/A]CAGACATGCCTGGGAGACAGAATAG
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
Gfod1 PubMed Links

Gene Details

Gene
Gfod1
Gene Name
glucose-fructose oxidoreductase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033399.4 Intron NP_001028571.1

View Full Product Details