Product Details

SNP ID
rs13478573
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:143752886 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTATATTTCCCAGGATGAACAGGGA[C/T]GTCTTCATGCCCCTCCTTAGCCTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
D130017N08Rik PubMed Links

Gene Details

Gene
D130017N08Rik
Gene Name
RIKEN cDNA D130017N08 gene
There are no transcripts associated with this gene.

Gene
Usp42
Gene Name
ubiquitin specific peptidase 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_029749.2 Intron NP_084025.2

View Full Product Details