Product Details

SNP ID
rs4178727
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:41878910 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCTCTATCAAATCCAGATAGCAGGA[A/G]TCTCTCTTCACCTCTGTCGCCCTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Lsamp PubMed Links

Gene Details

Gene
Lsamp
Gene Name
limbic system-associated membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175548.3 Intron NP_780757.1

View Full Product Details