Product Details

SNP ID
rs13483204
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:10544740 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTACACATGAACAGGCACATCTTGA[C/T]TTTTAAACTTGCTTGCTAGGGCTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Greb1l PubMed Links

Gene Details

Gene
Greb1l
Gene Name
growth regulation by estrogen in breast cancer-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083628.1 Intron NP_001077097.1

View Full Product Details