Product Details

SNP ID
rs13459119
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:20325683 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GTCCCCCTCTACACAAGAGTCTGGA[C/T]GCACGAGAAAAGTCCATCTTCAGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Bclaf1 PubMed Links

Gene Details

Gene
Bclaf1
Gene Name
BCL2-associated transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025392.1 1798 Silent Mutation NP_001020563.1
NM_001025393.1 1798 Silent Mutation NP_001020564.1
NM_153787.2 1798 Silent Mutation NP_722482.1

View Full Product Details