Product Details

SNP ID
rs6175578
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:40628378 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGTTTTCCTGGGATTAGAATC[C/T]ACCTTTCTAGCATACATGGTTTTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Lrp1b PubMed Links

Gene Details

Gene
Lrp1b
Gene Name
low density lipoprotein-related protein 1B (deleted in tumors)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053011.2 Intron NP_443737.2

View Full Product Details