Product Details
- SNP ID
-
rs3022887
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:69083134 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTGGGTGTTTCTATGATAACGCTC[G/A]TCTCCCCCTTTCAGGGTGATAGACA
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Cers6
PubMed Links
Gene Details
- Gene
- Cers6
- Gene Name
- ceramide synthase 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172856.3 |
|
Intron |
|
|
NP_766444.1 |
View Full Product Details