Product Details
- SNP ID
-
rs6412631
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:77216266 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGTCCTGACCTATTGTCTTATAAG[A/T]ATGTGGGGATACAATTAAACTGGCT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Sestd1
PubMed Links
Gene Details
- Gene
- Sestd1
- Gene Name
- SEC14 and spectrin domains 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175465.6 |
|
Intron |
|
|
NP_780674.1 |
View Full Product Details