Product Details
- SNP ID
-
rs6343480
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:192105460 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCCATTTTGAAATAAGAGTATTGG[C/T]ATCTAAAAACTGTACGGATATGGCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Rcor3
PubMed Links
Gene Details
- Gene
- Rcor3
- Gene Name
- REST corepressor 3
There are no transcripts associated with this gene.
View Full Product Details