Product Details
- SNP ID
-
rs13464873
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:128258983 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTGAGACACCAGCTACCTCTGACA[C/T]AAAGTCAGATACTGCAACAGGTATA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ubxn4
PubMed Links
Gene Details
- Gene
- Ubxn4
- Gene Name
- UBX domain protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_026390.2 |
775 |
Nonsense Mutation |
|
|
NP_080666.2 |
View Full Product Details