Product Details

SNP ID
rs13476110
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:131130075 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGCACTTCACCGCGCTCAGGCTTTC[C/T]GAACTTTTCCCCTGAGAAGCCGGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Dyrk3 PubMed Links

Gene Details

Gene
Dyrk3
Gene Name
dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145508.2 438 Silent Mutation NP_663483.1

View Full Product Details