Product Details
- SNP ID
-
rs13481383
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:33061769 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACCATGGTTCCTAGAGGCACTTC[C/T]TCTGGGACAGAGTACACTCTCCTGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Cdhr3
PubMed Links
Gene Details
- Gene
- Cdhr3
- Gene Name
- cadherin-related family member 3
View Full Product Details