Product Details
- SNP ID
-
rs13481106
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:75906610 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTGGGAGAGACCGTTGCCCATCA[C/T]GTTCCTCTGCATTGTCTCCAGCCGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Rph3al
PubMed Links
Gene Details
- Gene
- Rph3al
- Gene Name
- rabphilin 3A-like (without C2 domains)
There are no transcripts associated with this gene.
View Full Product Details