Product Details
- SNP ID
-
rs13483913
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:101102113 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACATTCATTTCCTTCATGAGGCAG[G/T]TCTACTCAAAGGCCAAGATCATTAG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Snx12
PubMed Links
Gene Details
- Gene
- Snx12
- Gene Name
- sorting nexin 12
View Full Product Details