Product Details

SNP ID
rs13466184
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:87718792 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATTCATGGCTGAAATGCTGGAGACT[G/T]CTTCCATCCTCAGGTATGTGTCCTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Msh2 PubMed Links

Gene Details

Gene
Msh2
Gene Name
mutS homolog 2 (E. coli)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008628.2 2236 Missense Mutation NP_032654.1

View Full Product Details