Product Details

SNP ID
rs3666799
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:31887673 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATCATCTTTATGCGTCTGTAAAATT[T/G]AACTTTCTCTCTTTCTCCCTAAGGC
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
Wdr33 PubMed Links

Gene Details

Gene
Wdr33
Gene Name
WD repeat domain 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170966.1 Intron NP_001164437.1
NM_001170967.1 Intron NP_001164438.1
NM_001170970.1 Intron NP_001164441.1
NM_028866.3 Intron NP_083142.2

View Full Product Details