Product Details
- SNP ID
-
rs13482077
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:16959996 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTCTCTCACTGCTGGAGAAGGGA[A/G]ACTCAGCCACTGATTGTATGTGCAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Rarb
PubMed Links
Gene Details
- Gene
- Rarb
- Gene Name
- retinoic acid receptor, beta
There are no transcripts associated with this gene.
View Full Product Details