Product Details

SNP ID
rs13482644
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:72628442 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCAAGGCACAATCTCTGCATCAGGA[C/T]ACACGCATGGGCAAGGACACCAAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Trappc9 PubMed Links

Gene Details

Gene
Trappc9
Gene Name
trafficking protein particle complex 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164641.1 Intron NP_001158113.1
NM_001164642.1 Intron NP_001158114.1
NM_001164643.1 Intron NP_001158115.1
NM_029640.2 Intron NP_083916.1
NM_180662.2 Intron NP_850993.2

View Full Product Details