Product Details

SNP ID
rs13482569
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:37780705 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAACTGAGAGACAGGCATCAATCCA[C/T]AGGGAGCCACTCTGAATGCAAGAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Ncald PubMed Links

Gene Details

Gene
Ncald
Gene Name
neurocalcin delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170866.1 Intron NP_001164337.1
NM_001170867.1 Intron NP_001164338.1
NM_001170868.1 Intron NP_001164339.1
NM_134094.4 Intron NP_598855.2

View Full Product Details