Product Details

SNP ID
rs3023778
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:51739714 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CACCCGGGGCTGCCAAGCAGTTTTG[G/T]AAAACACAGTAAAACCTTGAGTGCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Satb1 PubMed Links

Gene Details

Gene
Satb1
Gene Name
special AT-rich sequence binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163630.1 2641 UTR 3 NP_001157102.1
NM_001163631.1 2641 UTR 3 NP_001157103.1
NM_001163632.1 2641 UTR 3 NP_001157104.1
NM_009122.2 2641 UTR 3 NP_033148.2

View Full Product Details