Product Details
- SNP ID
-
rs4222308
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:40856560 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAACACTCCAGGGGTTTAGCTCAA[C/T]TCACAATCACAAACCATTTCAAGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Tmem182
PubMed Links
Gene Details
- Gene
- Tmem182
- Gene Name
- transmembrane protein 182
View Full Product Details