Product Details
- SNP ID
-
rs4229416
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:73043334 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAAGGGTATTAAGAGTTGGAGAGG[A/G]GGGAGAGGATGCTCAACTGAGACCG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Six1
PubMed Links
Gene Details
- Gene
- Six1
- Gene Name
- sine oculis-related homeobox 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_009189.3 |
1342 |
UTR 3 |
|
|
NP_033215.2 |
View Full Product Details