Product Details

SNP ID
rs13477685
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:47371402 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TAGAAAGAAAAGTTTATTTGGGAAA[A/T]TAGAACCTTCTATGGCTATTTCTTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Tgfbr1 PubMed Links

Gene Details

Gene
Tgfbr1
Gene Name
transforming growth factor, beta receptor I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_009370.2 Intron NP_033396.1

View Full Product Details