Product Details

SNP ID
rs13483376
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:56786096 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTATAGTAAAAATATTTGAAAATTT[A/C]AATAGCAAGACATTACTTAGGTTCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
March3 PubMed Links

Gene Details

Gene
March3
Gene Name
membrane-associated ring finger (C3HC4) 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177115.2 Intron NP_796089.1

View Full Product Details