Product Details
- SNP ID
-
rs13482051
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:9623813 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAAGTCACAAGTGAAGACAGATGT[C/T]CCACTAAGTAATGAACTGAGAGCAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Fhit
PubMed Links
Gene Details
- Gene
- Fhit
- Gene Name
- fragile histidine triad gene
There are no transcripts associated with this gene.
View Full Product Details