Product Details

SNP ID
rs4217517
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:90484481 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCTCCACACAGTAGCCAGAGT[A/G]GGCTTGTCCCTATCCTTCTTAATGC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
Hunk PubMed Links

Gene Details

Gene
Hunk
Gene Name
hormonally upregulated Neu-associated kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015755.2 Intron NP_056570.1

View Full Product Details