Product Details
- SNP ID
-
rs4228477
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:115078835 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATCCCTTTGTTTCTCTACGACTCC[A/G]TCTCCAACTCTTTGGAATCAGAGAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Tph2
PubMed Links
Gene Details
- Gene
- Tph2
- Gene Name
- tryptophan hydroxylase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173391.3 |
2432 |
UTR 3 |
|
|
NP_775567.2 |
View Full Product Details