Product Details
- SNP ID
-
rs13480951
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:33638734 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTAAATCTAACTTCCTGGCATTT[C/T]GTTTAGAATTTGTGACACATATTTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Kcnip1
PubMed Links
Gene Details
- Gene
- Kcnip1
- Gene Name
- Kv channel-interacting protein 1
View Full Product Details