Product Details
- SNP ID
-
rs13482434
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:12164234 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAATAATTGAGAATTGTACTTGAC[C/T]ACAGTATAGCAACACAAAACATAAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Mir1898
PubMed Links
Gene Details
- Gene
- Mir1898
- Gene Name
- microRNA 1898
There are no transcripts associated with this gene.
- Gene
- Zfr
- Gene Name
- zinc finger RNA binding protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_011767.2 |
|
Intron |
|
|
NP_035897.2 |
View Full Product Details