Product Details
- SNP ID
-
rs13482429
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:10575512 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGGGTTTCCTTGAGCTTCTGACT[C/G]AGGTCAGAGCCATTCTCGGAGATCT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Rai14
PubMed Links
Gene Details
- Gene
- Rai14
- Gene Name
- retinoic acid induced 14
View Full Product Details