Product Details

SNP ID
rs13483172
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:93202098 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGTGTAGTTCCCTTACCTTGTTCCC[A/G]GGACCATTCTGGAAGTAAGGGCAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Adcyap1 PubMed Links

Gene Details

Gene
Adcyap1
Gene Name
adenylate cyclase activating polypeptide 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_009625.2 Intron NP_033755.1

View Full Product Details