Product Details
- SNP ID
-
rs13481555
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:80348923 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAAAATAGGATAACACAAAGAAC[C/T]CTTCCTTTGCCCTTGGACAGCAGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Dcaf5
PubMed Links
Gene Details
- Gene
- Dcaf5
- Gene Name
- DDB1 and CUL4 associated factor 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177267.4 |
|
Intron |
|
|
NP_796241.3 |
View Full Product Details