Product Details

SNP ID
rs13481056
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:62265001 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAGGCCCGTGGGACACCAGCAGGGG[C/T]GGGCCTCGGGTCAAGGGATGCTGTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Adora2b PubMed Links

Gene Details

Gene
Adora2b
Gene Name
adenosine A2b receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007413.4 Intron NP_031439.2
Gene
Zswim7
Gene Name
zinc finger SWIM-type containing 7
There are no transcripts associated with this gene.

View Full Product Details