Product Details

SNP ID
rs6283329
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:23028398 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGCCACCTTTAGAGGTCTGCACATC[A/T]AACTTGGGATATGCACGCATTGCCT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Fam161a PubMed Links

Gene Details

Gene
Fam161a
Gene Name
family with sequence similarity 161, member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_028672.2 Intron NP_082948.2

View Full Product Details