Product Details
- SNP ID
-
rs13477669
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:46642822 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTCCTGAGAAGTGGGCGGTGTGC[A/C]TAGAAAGCCATCAGCTACTGAAGAG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Tbc1d2
PubMed Links
Gene Details
- Gene
- Tbc1d2
- Gene Name
- TBC1 domain family, member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198664.3 |
|
Intron |
|
|
NP_941066.3 |
View Full Product Details