Product Details
- SNP ID
-
rs30193415
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:55930069 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCAGTAGATAAACCATTTGTTAG[A/G]TGCTCTCAAGGTCTAATTATGCCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ccdc129
PubMed Links
Gene Details
- Gene
- Ccdc129
- Gene Name
- coiled-coil domain containing 129
View Full Product Details