Product Details

SNP ID
rs13483074
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:65610503 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AACACACACTTGGAATTCTTTTTTT[A/G]CAAACTTTCTTCTTCAAACTTCCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Vapa PubMed Links

Gene Details

Gene
Vapa
Gene Name
vesicle-associated membrane protein, associated protein A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013933.3 Intron NP_038961.2

View Full Product Details