Product Details
- SNP ID
-
rs13481552
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:79385348 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACCTCAAGGCATTTCAGAGCTGCC[A/G]CTGGAGGCTCCGTGATCTTCCCAGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Rad51b
PubMed Links
Gene Details
- Gene
- Rad51b
- Gene Name
- RAD51 homolog B
View Full Product Details