Product Details
- SNP ID
-
rs13480613
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:50969467 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAGAAATCTTTGTTAGGTTGGTT[A/C]CAGGCACATTTAATAAACTAAATCC
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Sim1
PubMed Links
Gene Details
- Gene
- Sim1
- Gene Name
- single-minded homolog 1 (Drosophila)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_011376.3 |
|
Intron |
|
|
NP_035506.2 |
View Full Product Details